missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,2,3,3-Tetramethylcyclopropanecarboxylic Acid, TCI America™

Manufacturer:  TCI America T23505G

 View more versions of this product

Catalog No. T23505G



Specifications

Specifications

2,2,3,3-Tetramethylcyclopropanecarboxylic Acid
120°C
15641-58-4
MFCD00011538
SFHVXKNMCGSLAR-UHFFFAOYSA-N
2,2,3,3-tetramethylcyclopropane-1-carboxylic acid
152115
142.20
≥98.0% (GC,T)
White
5g
C8H14O2
2,2,3,3-tetramethylcyclopropanecarboxylic acid, tetramethylcyclopropane-1-carboxylic acid, cyclopropanecarboxylic acid, 2,2,3,3-tetramethyl, 2,2,3,3-tetramethylcyclopropanecarboxylicacid, acmc-1c48r, tetramethylcyclopropanecarboxylicacid, 3-carboxy-1,1,2,2-tetramethylcyclopropane, 2,2,3,3-tetramethyl cyclopropanecarboxylic acid, 2,2,3,3-tetramethyl-cyclopropanecarboxylic acid, 2,2,3,3-tetramethylcyclopropane carboxylic acid
CC1(C(C1(C)C)C(=O)O)C
142.198
CHEBI:39355
Crystal-Powder at 20°C

Chemical Identifiers

15641-58-4
142.198
SFHVXKNMCGSLAR-UHFFFAOYSA-N
152115
2,2,3,3-tetramethylcyclopropane-1-carboxylic acid
C8H14O2
MFCD00011538
2,2,3,3-tetramethylcyclopropanecarboxylic acid, tetramethylcyclopropane-1-carboxylic acid, cyclopropanecarboxylic acid, 2,2,3,3-tetramethyl, 2,2,3,3-tetramethylcyclopropanecarboxylicacid, acmc-1c48r, tetramethylcyclopropanecarboxylicacid, 3-carboxy-1,1,2,2-tetramethylcyclopropane, 2,2,3,3-tetramethyl cyclopropanecarboxylic acid, 2,2,3,3-tetramethyl-cyclopropanecarboxylic acid, 2,2,3,3-tetramethylcyclopropane carboxylic acid
CHEBI:39355
CC1(C(C1(C)C)C(=O)O)C
Safety and Handling

Safety and Handling

TSCA : No

Documents