missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,2,3,3-Tetramethylcyclopropanecarboxylic acid, 97%

$44.72 - $178.89

Chemical Identifiers

CAS 15641-58-4
Molecular Formula C8H14O2
Molecular Weight (g/mol) 142.198
MDL Number MFCD00011538
InChI Key SFHVXKNMCGSLAR-UHFFFAOYSA-N
Synonym 2,2,3,3-tetramethylcyclopropanecarboxylic acid, tetramethylcyclopropane-1-carboxylic acid, cyclopropanecarboxylic acid, 2,2,3,3-tetramethyl, 2,2,3,3-tetramethylcyclopropanecarboxylicacid, acmc-1c48r, tetramethylcyclopropanecarboxylicacid, 3-carboxy-1,1,2,2-tetramethylcyclopropane, 2,2,3,3-tetramethyl cyclopropanecarboxylic acid, 2,2,3,3-tetramethyl-cyclopropanecarboxylic acid, 2,2,3,3-tetramethylcyclopropane carboxylic acid
PubChem CID 152115
ChEBI CHEBI:39355
IUPAC Name 2,2,3,3-tetramethylcyclopropane-1-carboxylic acid
SMILES CC1(C(C1(C)C)C(=O)O)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6080903 View Documents Alfa Aesar™
H6080903
1g Each for $44.72
 
AAH6080906 View Documents Alfa Aesar™
H6080906
5g Each for $178.89
 
Description

Description

Applications
2,2,3,3-Tetramethylcyclopropanecarboxylic acid is used to start synthesis of central nervous system-active compounds.

Solubility
Slightly soluble in water. Soluble in methanol, ethanol and acetonitrile.

Notes
Keep container tightly sealed. Store in refrigerated conditions. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

15641-58-4
142.198
SFHVXKNMCGSLAR-UHFFFAOYSA-N
152115
2,2,3,3-tetramethylcyclopropane-1-carboxylic acid
C8H14O2
MFCD00011538
2,2,3,3-tetramethylcyclopropanecarboxylic acid, tetramethylcyclopropane-1-carboxylic acid, cyclopropanecarboxylic acid, 2,2,3,3-tetramethyl, 2,2,3,3-tetramethylcyclopropanecarboxylicacid, acmc-1c48r, tetramethylcyclopropanecarboxylicacid, 3-carboxy-1,1,2,2-tetramethylcyclopropane, 2,2,3,3-tetramethyl cyclopropanecarboxylic acid, 2,2,3,3-tetramethyl-cyclopropanecarboxylic acid, 2,2,3,3-tetramethylcyclopropane carboxylic acid
CHEBI:39355
CC1(C(C1(C)C)C(=O)O)C

Specifications

120°C to 121°C
15641-58-4
MFCD00011538
SFHVXKNMCGSLAR-UHFFFAOYSA-N
2,2,3,3-tetramethylcyclopropane-1-carboxylic acid
152115
142.2
2,2,3,3-Tetramethylcyclopropanecarboxylic acid
1g
C8H14O2
2,2,3,3-tetramethylcyclopropanecarboxylic acid, tetramethylcyclopropane-1-carboxylic acid, cyclopropanecarboxylic acid, 2,2,3,3-tetramethyl, 2,2,3,3-tetramethylcyclopropanecarboxylicacid, acmc-1c48r, tetramethylcyclopropanecarboxylicacid, 3-carboxy-1,1,2,2-tetramethylcyclopropane, 2,2,3,3-tetramethyl cyclopropanecarboxylic acid, 2,2,3,3-tetramethyl-cyclopropanecarboxylic acid, 2,2,3,3-tetramethylcyclopropane carboxylic acid
CC1(C(C1(C)C)C(=O)O)C
142.198
CHEBI:39355
97%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : GZ2083500