Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,2,3,3-Tetrafluoropropyl methacrylate, 97%, stab. with 50ppm 4-methoxyphenol

Chemical Identifiers

Chemical Name or Material 2,2,3,3-Tetrafluoropropyl methacrylate
CAS 45102-52-1
Name Note stabilized with 50ppm 4-methoxyphenol
Molecular Formula C7H8F4O2
Formula Weight 200.13
MDL Number MFCD00042381
UN Number UN3272
Beilstein 3588831
Synonym Methacrylic acid 2,2,3,3-tetrafluoropropyl ester; 2,2,3,3-Tetrafluoropropyl 2-methylpropenoate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2268206

 
alfa aesar™
B2268206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2268214

 
alfa aesar™
B2268214
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 200.13
Chemical Name or Material 2,2,3,3-Tetrafluoropropyl methacrylate
Name Note stabilized with 50ppm 4-methoxyphenol
Assay 97%
Molecular Formula C7H8F4O2
MDL Number MFCD00042381
CAS 45102-52-1
UN Number UN3272
Beilstein 3588831
Synonym Methacrylic acid 2,2,3,3-tetrafluoropropyl ester; 2,2,3,3-Tetrafluoropropyl 2-methylpropenoate
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Boiling Point 146° to 149°C
Flash Point 50°C (122°F)
Quantity 5g
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
Safety and Handling

Warning

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.