missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,2,3,3-Tetrafluorocyclobutanol, 95%

Chemical Identifiers

CAS 374-32-3
Molecular Formula C4H4F4O
Formula Weight 144.07
MDL Number MFCD00190085
UN Number UN1987
Beilstein 2429286
Chemical Name or Material 2,2,3,3-Tetrafluorocyclobutanol
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1918603 SDS Alfa Aesar™
L1918603
1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAL1918606 SDS Alfa Aesar™
L1918606
5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Description

Applications
2,2,3,3-Tetrafluorocyclobutanol is used as a pharmaceutical intermediate.

Solubility
Not miscible or difficult to mix in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
Description & Specifications

Chemical Identifiers

374-32-3
144.07
UN1987
2,2,3,3-Tetrafluorocyclobutanol
C4H4F4O
MFCD00190085
2429286

Specifications

374-32-3
144.07
UN1987
155° to 157°C
1g
95%
C4H4F4O
MFCD00190085
2429286
44°C (111°F)
2,2,3,3-Tetrafluorocyclobutanol

Safety and Handling

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.