missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,2,3,3-Tetrafluoro-1,4-butanediol, TCI America™

Manufacturer:  TCI America T229625G

 View more versions of this product

Catalog No. T229625G



Specifications

Specifications

2,2,3,3-Tetrafluoro-1,4-butanediol
White
25g
C4H6F4O2
2,2,3,3-tetrafluoro-1,4-butanediol, 1,4-dihydroxy-2,2,3,3-tetrafluorobutane, acmc-1aqx7, 1,4-butanediol,2,2,3,3-tetrafluoro, 2,2,3,3-tetrakis fluoranyl butane-1,4-diol
C(C(C(CO)(F)F)(F)F)O
162.084
162.08
≥95.0% (GC)
112°C
80°C
425-61-6
MFCD00042375
CDZXJJOGDCLNKX-UHFFFAOYSA-N
2,2,3,3-tetrafluorobutane-1,4-diol
136270
Crystal-Powder at 20°C

Chemical Identifiers

425-61-6
162.084
CDZXJJOGDCLNKX-UHFFFAOYSA-N
136270
C(C(C(CO)(F)F)(F)F)O
C4H6F4O2
MFCD00042375
2,2,3,3-tetrafluoro-1,4-butanediol, 1,4-dihydroxy-2,2,3,3-tetrafluorobutane, acmc-1aqx7, 1,4-butanediol,2,2,3,3-tetrafluoro, 2,2,3,3-tetrakis fluoranyl butane-1,4-diol
2,2,3,3-tetrafluorobutane-1,4-diol
Safety and Handling

Safety and Handling

TSCA : No

Documents