missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2,2',3,3',5,5',6,6'-Octafluorobiphenyl 97.0+%, TCI America™
Supplier: TCI America O01821G
Specifications
2,2′,3,3′,5,5′,6,6′-Octafluorobiphenyl | |
1 g | |
C12H2F8 | |
4H,4′CH-Octafluorobiphenyl | |
FC1=CC(F)=C(F)C(=C1F)C1=C(F)C(F)=CC(F)=C1F | |
298.14 | |
298.14 | |
85°C |
3883-86-1 | |
White | |
MFCD00000312 | |
QWCHHUZAAGRHDB-UHFFFAOYSA-N | |
2,2',3,3',5,5',6,6'-octafluoro-1,1'-biphenyl | |
77492 | |
≥97.0% (GC) | |
Powder/Crystals |
Chemical Identifiers
3883-86-1 | |
298.14 | |
QWCHHUZAAGRHDB-UHFFFAOYSA-N | |
77492 | |
FC1=CC(F)=C(F)C(=C1F)C1=C(F)C(F)=CC(F)=C1F |
C12H2F8 | |
MFCD00000312 | |
4H,4′CH-Octafluorobiphenyl | |
2,2',3,3',5,5',6,6'-octafluoro-1,1'-biphenyl |
Safety and Handling
TSCA : Yes