missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,2,3,3,4,4-Hexafluoropentanedioic Anhydride, TCI America™

$35.14 - $229.17

Chemical Identifiers

CAS 376-68-1
Molecular Formula C5F6O3
Molecular Weight (g/mol) 222.042
MDL Number MFCD00006680
InChI Key IHYAGCYJVNHXCT-UHFFFAOYSA-N
Synonym hexafluoroglutaric anhydride, perfluoroglutaric anhydride, 3,3,4,4,5,5-hexafluorodihydro-2h-pyran-2,6 3h-dione, hexafluoroglutaric acid anhydride, 2,2,3,3,4,4-hexafluoropentanedioic anhydride, 3,3,4,4,5,5-hexafluoro-3h-4,5-dihydropyran-2,6-dione, hexafluoroglutaricanhydride, acmc-1clz4, hexafluorooxane-2,6-dione, hexafluoroglutaric anhydride
PubChem CID 78995
IUPAC Name 3,3,4,4,5,5-hexafluorooxane-2,6-dione
SMILES C1(=O)C(C(C(C(=O)O1)(F)F)(F)F)(F)F
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H07451G View Documents TCI America
H07451G
1g Each for $35.14
 
H074525G View Documents TCI America
H074525G
25g Each for $229.17
 
H07455G View Documents TCI America
H07455G
5g Each for $93.20
 
Specifications

Chemical Identifiers

376-68-1
222.042
IHYAGCYJVNHXCT-UHFFFAOYSA-N
78995
C1(=O)C(C(C(C(=O)O1)(F)F)(F)F)(F)F
C5F6O3
MFCD00006680
hexafluoroglutaric anhydride, perfluoroglutaric anhydride, 3,3,4,4,5,5-hexafluorodihydro-2h-pyran-2,6 3h-dione, hexafluoroglutaric acid anhydride, 2,2,3,3,4,4-hexafluoropentanedioic anhydride, 3,3,4,4,5,5-hexafluoro-3h-4,5-dihydropyran-2,6-dione, hexafluoroglutaricanhydride, acmc-1clz4, hexafluorooxane-2,6-dione, hexafluoroglutaric anhydride
3,3,4,4,5,5-hexafluorooxane-2,6-dione

Specifications

72°C
1g
C5F6O3
hexafluoroglutaric anhydride, perfluoroglutaric anhydride, 3,3,4,4,5,5-hexafluorodihydro-2h-pyran-2,6 3h-dione, hexafluoroglutaric acid anhydride, 2,2,3,3,4,4-hexafluoropentanedioic anhydride, 3,3,4,4,5,5-hexafluoro-3h-4,5-dihydropyran-2,6-dione, hexafluoroglutaricanhydride, acmc-1clz4, hexafluorooxane-2,6-dione, hexafluoroglutaric anhydride
C1(=O)C(C(C(C(=O)O1)(F)F)(F)F)(F)F
222.042
222.04
≥97.0% (T)
Yellow
376-68-1
MFCD00006680
IHYAGCYJVNHXCT-UHFFFAOYSA-N
3,3,4,4,5,5-hexafluorooxane-2,6-dione
78995
Clear Liquid at 20°C
2,2,3,3,4,4-Hexafluoropentanedioic Anhydride
Safety and Handling

Safety and Handling

RTECSNumber : TM0860000

TSCA : No