Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,2,3,3,4,4-Hexafluoro-1,5-pentanediol, 98%

$145.69 - $145.69

Chemical Identifiers

Chemical Name or Material 2,2,3,3,4,4-Hexafluoro-1,5-pentanediol
CAS 376-90-9
Molecular Formula HOCH2(CF2)3CH2OH
Formula Weight 212.09g/mol
MDL Number MFCD00042434
Beilstein 1766258
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2191706

 
alfa aesar™
B21917-06
5g Each for $145.69
Description & Specifications

Specifications

Boiling Point 110° to 112°C (10mmHg)
Melting Point 76° to 80°C
Quantity 5g
Flash Point >110°C (230°F)
Chemical Name or Material 2,2,3,3,4,4-Hexafluoro-1,5-pentanediol
CAS 376-90-9
Assay 98%
Molecular Formula HOCH2(CF2)3CH2OH
Formula Weight 212.09g/mol
MDL Number MFCD00042434
Beilstein 1766258
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number SA0750000
TSCA TSCA
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.