missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2,2,3,3,4,4,5,5-Octafluoropentyl methacrylate, 98%, stab., Thermo Scientific Chemicals
$145.56 - $484.15
Chemical Identifiers
CAS | 355-93-1 |
---|---|
Molecular Formula | C9H8F8O2 |
Molecular Weight (g/mol) | 300.15 |
MDL Number | MFCD00039278 |
InChI Key | ZNJXRXXJPIFFAO-UHFFFAOYSA-N |
Synonym | 1h,1h,5h-octafluoropentyl methacrylate, 2,2,3,3,4,4,5,5-octafluoropentyl methacrylate, octafluoropentyl methacrylate, 1h,1h,5h-perfluoropentyl methacrylate, 1h,1h,5h-octafluoropentyl methacrylate stabilized with mehq, 2-propenoic acid, 2-methyl-, 2,2,3,3,4,4,5,5-octafluoropentyl ester, methacrylic acid 1h,1h,5h-perfluoropentyl ester, methacrylic acid 1h,1h,5h-octafluoropentyl ester, octafluoropentyl methacrylate polymer, 1h,1h,5h-octafluoropentylmethacrylate |
PubChem CID | 67739 |
IUPAC Name | 2,2,3,3,4,4,5,5-octafluoropentyl 2-methylprop-2-enoate |
SMILES | CC(=C)C(=O)OCC(F)(F)C(F)(F)C(F)(F)C(F)F |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL1204406
|
Thermo Scientific Chemicals
L1204406 |
5 g |
Each for $145.56
|
|
|||||
AAL1204414
|
Thermo Scientific Chemicals
L1204414 |
25 g |
Each for $484.15
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
355-93-1 | |
300.15 | |
ZNJXRXXJPIFFAO-UHFFFAOYSA-N | |
67739 | |
CC(=C)C(=O)OCC(F)(F)C(F)(F)C(F)(F)C(F)F |
C9H8F8O2 | |
MFCD00039278 | |
1h,1h,5h-octafluoropentyl methacrylate, 2,2,3,3,4,4,5,5-octafluoropentyl methacrylate, octafluoropentyl methacrylate, 1h,1h,5h-perfluoropentyl methacrylate, 1h,1h,5h-octafluoropentyl methacrylate stabilized with mehq, 2-propenoic acid, 2-methyl-, 2,2,3,3,4,4,5,5-octafluoropentyl ester, methacrylic acid 1h,1h,5h-perfluoropentyl ester, methacrylic acid 1h,1h,5h-octafluoropentyl ester, octafluoropentyl methacrylate polymer, 1h,1h,5h-octafluoropentylmethacrylate | |
2,2,3,3,4,4,5,5-octafluoropentyl 2-methylprop-2-enoate |
Specifications
355-93-1 | |
178°C to 179°C | |
C9H8F8O2 | |
MFCD00039278 | |
1801409 | |
ZNJXRXXJPIFFAO-UHFFFAOYSA-N | |
2,2,3,3,4,4,5,5-octafluoropentyl 2-methylprop-2-enoate | |
67739 | |
98% |
1.432 | |
81°C (177°F) | |
1.36 | |
5 g | |
1h,1h,5h-octafluoropentyl methacrylate, 2,2,3,3,4,4,5,5-octafluoropentyl methacrylate, octafluoropentyl methacrylate, 1h,1h,5h-perfluoropentyl methacrylate, 1h,1h,5h-octafluoropentyl methacrylate stabilized with mehq, 2-propenoic acid, 2-methyl-, 2,2,3,3,4,4,5,5-octafluoropentyl ester, methacrylic acid 1h,1h,5h-perfluoropentyl ester, methacrylic acid 1h,1h,5h-octafluoropentyl ester, octafluoropentyl methacrylate polymer, 1h,1h,5h-octafluoropentylmethacrylate | |
CC(=C)C(=O)OCC(F)(F)C(F)(F)C(F)(F)C(F)F | |
300.15 | |
300.15 | |
2,2,3,3,4,4,5,5-Octafluoropentyl methacrylate, stabilized |
Safety and Handling
GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
P210-P280-P370+P378q-P501c
H227
EINECSNumber : 206-596-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only