Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,2,3,3,4,4,5,5-Octafluoropentyl acrylate, 97%, stab. with ca 50ppm 4-methoxyphenol

$60.80 - $189.34

Chemical Identifiers

Chemical Name or Material 2,2,3,3,4,4,5,5-Octafluoropentyl acrylate
CAS 376-84-1
Name Note stabilized with ≈50ppm 4-methoxyphenol
Molecular Formula C8F8H6O2
Linear Formula H2C=CHCO2CH2(CF2)4H
Formula Weight 286.12
MDL Number MFCD00039279
Synonym Acrylic acid 2,2,3,3,4,4,5,5-octafluoropentyl ester; 1H,1H,5H-Perfluoro-1-pentyl acrylate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1656106

  SDS
alfa aesar™
L1656106
5g Each for $60.80

AAL1656114

  SDS
alfa aesar™
L1656114
25g Each for $189.34
Description & Specifications

Specifications

Formula Weight 286.12
Chemical Name or Material 2,2,3,3,4,4,5,5-Octafluoropentyl acrylate
Assay 97%
Molecular Formula C8F8H6O2
Linear Formula H2C=CHCO2CH2(CF2)4H
MDL Number MFCD00039279
CAS 376-84-1
Synonym Acrylic acid 2,2,3,3,4,4,5,5-octafluoropentyl ester; 1H,1H,5H-Perfluoro-1-pentyl acrylate
Health Hazard 2 GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Flash Point 71°C (159°F)
Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage 2° to 8°C
Name Note stabilized with ≈50ppm 4-methoxyphenol
Health Hazard 1 Warning
Boiling Point 56°C (8mmHg)
Quantity 5g
RTECS Number UD3643645
Safety and Handling

Warning

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

2° to 8°C