Please login to your online account to display your discounted pricing

2,2,3,3,4,4,5,5-Octafluoro-1-pentanol, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2,2,3,3,4,4,5,5-Octafluoro-1-pentanol
Name Note 98%
CAS 355-80-6
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C5H4F8O
Linear Formula F2CH(CF2)3CH2OH
Formula Weight 232.07
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC312311000

  SDS
thermo fisher scientific
312311000
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Liquid
Formula Weight 232.07
Quantity 100g
Color Clear colorless
EINECS Number 206-593-4
TSCA TSCA
Density 1.6670g/mL
RTECS Number SA7650000
CAS Max % 100.0
Chemical Name or Material 2,2,3,3,4,4,5,5-Octafluoro-1-pentanol
Name Note 98%
Infrared Spectrum Authentic
Assay Percent Range 98%
Refractive Index 1.3170 to 1.3190 (20°C, 589nm)
Molecular Formula C5H4F8O
Linear Formula F2CH(CF2)3CH2OH
MDL Number MFCD00039631
CAS 355-80-6
CAS Min % 97.5
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.