missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,2,3,3,4,4,5,5-Octafluoro-1,6-hexanediol 98.0+%, TCI America™

Manufacturer:  TCI America O02945G

 View more versions of this product

Catalog No. O02945G



Specifications

Specifications

2,2,3,3,4,4,5,5-Octafluoro-1,6-hexanediol
68°C
355-74-8
MFCD00069087
NHEKBXPLFJSSBZ-UHFFFAOYSA-N
2,2,3,3,4,4,5,5-octafluorohexane-1,6-diol
136181
Crystal-Powder at 20°C
Undesignated
5g
C6H6F8O2
1,6-dihydroxy-2,2,3,3,4,4,5,5-octafluorohexane
C(C(C(C(C(CO)(F)F)(F)F)(F)F)(F)F)O
262.099
262.10
≥98.0% (GC)

Chemical Identifiers

355-74-8
262.099
NHEKBXPLFJSSBZ-UHFFFAOYSA-N
136181
C(C(C(C(C(CO)(F)F)(F)F)(F)F)(F)F)O
C6H6F8O2
MFCD00069087
1,6-dihydroxy-2,2,3,3,4,4,5,5-octafluorohexane
2,2,3,3,4,4,5,5-octafluorohexane-1,6-diol
Safety and Handling

Safety and Handling

TSCA : No

Documents