missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2,2,3,3,4,4,5,5,5-Nonafluoropentyloxirane 98.0+%, TCI America™
$31.52 - $350.05
Chemical Identifiers
CAS | 81190-28-5 |
---|---|
Molecular Formula | C7H5F9O |
Molecular Weight (g/mol) | 276.102 |
MDL Number | MFCD00236085 |
InChI Key | WUKHWLIEBSRTRH-UHFFFAOYSA-N |
Synonym | 3-Perfluorobutyl-1,2-epoxypropane |
PubChem CID | 2775819 |
IUPAC Name | 2-(2,2,3,3,4,4,5,5,5-nonafluoropentyl)oxirane |
SMILES | C1C(O1)CC(C(C(C(F)(F)F)(F)F)(F)F)(F)F |
Chemical Identifiers
81190-28-5 | |
276.102 | |
WUKHWLIEBSRTRH-UHFFFAOYSA-N | |
2775819 | |
C1C(O1)CC(C(C(C(F)(F)F)(F)F)(F)F)(F)F |
C7H5F9O | |
MFCD00236085 | |
3-Perfluorobutyl-1,2-epoxypropane | |
2-(2,2,3,3,4,4,5,5,5-nonafluoropentyl)oxirane |
Specifications
81190-28-5 | |
132°C | |
MFCD00236085 | |
1993 | |
WUKHWLIEBSRTRH-UHFFFAOYSA-N | |
2-(2,2,3,3,4,4,5,5,5-nonafluoropentyl)oxirane | |
2775819 | |
≥98.0% (GC) | |
2,2,3,3,4,4,5,5,5-Nonafluoropentyloxirane |
Colorless | |
C7H5F9O | |
5 g | |
3-Perfluorobutyl-1,2-epoxypropane | |
C1C(O1)CC(C(C(C(F)(F)F)(F)F)(F)F)(F)F | |
276.102 | |
276.10 | |
Liquid |
Safety and Handling
TSCA : No