Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,2,3,3,4,4,4-Heptafluorobutyl methacrylate, 97%, stab.

Chemical Identifiers

Chemical Name or Material 2,2,3,3,4,4,4-Heptafluorobutyl methacrylate
CAS 13695-31-3
Name Note stabilized
Molecular Formula C8H7F7O2
Formula Weight 268.13
MDL Number MFCD00039251
UN Number UN3272
Synonym Methacrylic acid 2,2,3,3,4,4,4-heptafluorobutyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1292206

  SDS
alfa aesar™
L1292206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1292214

  SDS
alfa aesar™
L1292214
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 268.13
Molecular Formula C8H7F7O2
Chemical Name or Material 2,2,3,3,4,4,4-Heptafluorobutyl methacrylate
Health Hazard 2 GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
MDL Number MFCD00039251
Assay 97%
Recommended Storage 2° to 8°C
Synonym Methacrylic acid 2,2,3,3,4,4,4-heptafluorobutyl ester
CAS 13695-31-3
Health Hazard 3 GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Warning
Quantity 5g
Name Note stabilized
TSCA TSCA
Boiling Point 134° to 136°C
UN Number UN3272
Odor Recognizable
Flash Point 36°C (96°F)
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.

Applications
2,2,3,3,4,4,4-Heptafluorobutyl methacrylate is used as an organic chemical synthesis intermediate.

Solubility
Not miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.

Safety and Handling

Warning

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C