missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,2,3,3,4,4,4-Heptafluorobutyl methacrylate, 97%, stab.

Manufacturer:  Alfa Aesar™ L1292206

 View more versions of this product

Catalog No. AAL1292206

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Applications
2,2,3,3,4,4,4-Heptafluorobutyl methacrylate is used as an organic chemical synthesis intermediate.

Solubility
Not miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.
Specifications

Specifications

2,2,3,3,4,4,4-Heptafluorobutyl methacrylate
Recognizable
5g
13695-31-3
MFCD00039251
heptafluorobutyl methacrylate, 2,2,3,3,4,4,4-heptafluorobutyl methacrylate, 1h,1h-heptafluorobutyl methacrylate, 1h,1h-heptafluoro-n-butyl methacrylate, 2-propenoic acid, 2-methyl-, 2,2,3,3,4,4,4-heptafluorobutyl ester, methacrylic acid, 2,2,3,3,4,4,4-heptafluorobutyl ester, methacrylic acid 2,2,3,3,4,4,4-heptafluorobutyl ester, heptafluoropropylmethyl methacrylate, 2,2,3,3,4,4,4-heptafluorobutyl 2-methylacrylate #, 2,2,3,3,4,4,4-heptafluorobutyl methacrylate
CC(=C)C(=O)OCC(C(C(F)(F)F)(F)F)(F)F
268.131
268.13
36°C (96°F)
134°C to 136°C
stabilized
C8H7F7O2
UN3272
VIEHKBXCWMMOOU-UHFFFAOYSA-N
2,2,3,3,4,4,4-heptafluorobutyl 2-methylprop-2-enoate
83666
97%

Chemical Identifiers

13695-31-3
268.131
VIEHKBXCWMMOOU-UHFFFAOYSA-N
83666
CC(=C)C(=O)OCC(C(C(F)(F)F)(F)F)(F)F
C8H7F7O2
MFCD00039251
heptafluorobutyl methacrylate, 2,2,3,3,4,4,4-heptafluorobutyl methacrylate, 1h,1h-heptafluorobutyl methacrylate, 1h,1h-heptafluoro-n-butyl methacrylate, 2-propenoic acid, 2-methyl-, 2,2,3,3,4,4,4-heptafluorobutyl ester, methacrylic acid, 2,2,3,3,4,4,4-heptafluorobutyl ester, methacrylic acid 2,2,3,3,4,4,4-heptafluorobutyl ester, heptafluoropropylmethyl methacrylate, 2,2,3,3,4,4,4-heptafluorobutyl 2-methylacrylate #, 2,2,3,3,4,4,4-heptafluorobutyl methacrylate
2,2,3,3,4,4,4-heptafluorobutyl 2-methylprop-2-enoate
Safety and Handling

Safety and Handling

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.

TSCA : TSCA

Recommended Storage : 2° to 8°C

Documents