missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Alfa Aesar™ 2,2,3,3,4,4,4-Heptafluorobutyl acrylate, 97%, stab. with 100 ppm 4-methoxyphenol
$68.59 - $220.78
Chemical Identifiers
CAS | 424-64-6 |
---|---|
Molecular Formula | C7H5F7O2 |
Molecular Weight (g/mol) | 254.104 |
MDL Number | MFCD00039252 |
InChI Key | PLXOUIVCSUBZIX-UHFFFAOYSA-N |
Synonym | 1,1-dihydroheptafluorobutyl acrylate, 1h, 1h-heptafluorobutylacrylate, 1h,1h-heptafluorobut-1-yl acrylat, 1h,1h-heptafluorobutyl acrylate, 2,2,3,3,4,4,4-heptafluorobutyl acrylate, 2,2,3,3,4,4,4-heptafluorobutyl acrylate, stab. with 100 ppm 4-methoxyphenol, 2,2,3,3,4,4,4-heptafluorobutyl=propenoate, 2-propenoic acid, 2,2,3,3,4,4,4-heptafluorobutyl ester, heptafluorobutyl acrylate |
PubChem CID | 67923 |
IUPAC Name | 2,2,3,3,4,4,4-heptafluorobutyl prop-2-enoate |
SMILES | C=CC(=O)OCC(C(C(F)(F)F)(F)F)(F)F |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL1040006
|
Alfa Aesar™
L1040006 |
5g |
Each for $68.59
|
|
AAL1040014
|
Alfa Aesar™
L1040014 |
25g |
Each for $220.78
|
|
Description
Applications2,2,3,3,4,4,4-Heptafluorobutyl acrylate is a useful fluorinated acrylate for proteomics research. It is also used as pharmaceutical intermediate.
Solubility
Not miscible in water.
Notes
Light sensitive. Incompatible with oxidizing agents, heat, ultraviolet radiation and free radical initiators.
Chemical Identifiers
424-64-6 | |
254.104 | |
PLXOUIVCSUBZIX-UHFFFAOYSA-N | |
67923 | |
C=CC(=O)OCC(C(C(F)(F)F)(F)F)(F)F |
C7H5F7O2 | |
MFCD00039252 | |
1,1-dihydroheptafluorobutyl acrylate, 1h, 1h-heptafluorobutylacrylate, 1h,1h-heptafluorobut-1-yl acrylat, 1h,1h-heptafluorobutyl acrylate, 2,2,3,3,4,4,4-heptafluorobutyl acrylate, 2,2,3,3,4,4,4-heptafluorobutyl acrylate, stab. with 100 ppm 4-methoxyphenol, 2,2,3,3,4,4,4-heptafluorobutyl=propenoate, 2-propenoic acid, 2,2,3,3,4,4,4-heptafluorobutyl ester, heptafluorobutyl acrylate | |
2,2,3,3,4,4,4-heptafluorobutyl prop-2-enoate |
Specifications
1.418 | |
Pungent | |
57°C to 58°C (30mmHg) | |
424-64-6 | |
MFCD00039252 | |
1792520 | |
Not miscible in water. | |
C=CC(=O)OCC(C(C(F)(F)F)(F)F)(F)F | |
254.104 | |
254.11 | |
2,2,3,3,4,4,4-Heptafluorobutyl acrylate, stabilized with 100ppm 4-methoxyphenol |
31°C (87°F) | |
1.331 | |
5g | |
C7H5F7O2 | |
UN3272 | |
1,1-dihydroheptafluorobutyl acrylate, 1h, 1h-heptafluorobutylacrylate, 1h,1h-heptafluorobut-1-yl acrylat, 1h,1h-heptafluorobutyl acrylate, 2,2,3,3,4,4,4-heptafluorobutyl acrylate, 2,2,3,3,4,4,4-heptafluorobutyl acrylate, stab. with 100 ppm 4-methoxyphenol, 2,2,3,3,4,4,4-heptafluorobutyl=propenoate, 2-propenoic acid, 2,2,3,3,4,4,4-heptafluorobutyl ester, heptafluorobutyl acrylate | |
PLXOUIVCSUBZIX-UHFFFAOYSA-N | |
2,2,3,3,4,4,4-heptafluorobutyl prop-2-enoate | |
67923 | |
97% |
Safety and Handling
GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c
H226-H302-H312-H315-H319-H332-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
EINECSNumber : 207-036-8
TSCA : Yes
Recommended Storage : Ambient temperatures