missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,2,3,3,4,4,4-Heptafluoro-1-butanol, 98%

$83.14 - $1,156.59

Chemical Identifiers

CAS 375-01-9
Molecular Formula C4H3F7O
Molecular Weight (g/mol) 200.056
MDL Number MFCD00004674
InChI Key WXJFKAZDSQLPBX-UHFFFAOYSA-N
Synonym 2,2,3,3,4,4,4-heptafluoro-1-butanol, 1-butanol, 2,2,3,3,4,4,4-heptafluoro, 1h,1h-heptafluorobutanol-1, 1h,1h-heptafluoro-1-butanol, 1,1-dihydroperfluorobutyl alcohol, 2,2,3,3,4,4,4-heptafluorobutanol, 1,1-dihydroperfluorobutanol, 1,1-h,h-heptafluorobutanol, perfluoro-1,1-dihydrobutanol, butanol, 2,2,3,3,4,4,4-heptafluoro
PubChem CID 9776
IUPAC Name 2,2,3,3,4,4,4-heptafluorobutan-1-ol
SMILES C(C(C(C(F)(F)F)(F)F)(F)F)O
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1041430 View Documents Alfa Aesar™
A1041430
250g Each for $1,156.59
 
AAA1041414 View Documents Alfa Aesar™
A1041414
25g Each for $168.09
 
AAA1041409 View Documents Alfa Aesar™
A1041409
10g Each for $83.14
 
AAA1041418 View Documents Alfa Aesar™
A1041418
50g Each for $296.08
 
Description

Description

Applications
It is employed as a halogenated solvent in electrospray liquid chromatography.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Protect from electrostatic charges and keep away from ignition sources.
Specifications

Chemical Identifiers

375-01-9
200.056
WXJFKAZDSQLPBX-UHFFFAOYSA-N
9776
C(C(C(C(F)(F)F)(F)F)(F)F)O
C4H3F7O
MFCD00004674
2,2,3,3,4,4,4-heptafluoro-1-butanol, 1-butanol, 2,2,3,3,4,4,4-heptafluoro, 1h,1h-heptafluorobutanol-1, 1h,1h-heptafluoro-1-butanol, 1,1-dihydroperfluorobutyl alcohol, 2,2,3,3,4,4,4-heptafluorobutanol, 1,1-dihydroperfluorobutanol, 1,1-h,h-heptafluorobutanol, perfluoro-1,1-dihydrobutanol, butanol, 2,2,3,3,4,4,4-heptafluoro
2,2,3,3,4,4,4-heptafluorobutan-1-ol

Specifications

25°C (77°F)
250g
C4H3F7O
MFCD00004674
1761907
WXJFKAZDSQLPBX-UHFFFAOYSA-N
2,2,3,3,4,4,4-heptafluorobutan-1-ol
9776
98%
95°C to 97°C
375-01-9
CF3CF2CF2CH2OH
UN1987
2,2,3,3,4,4,4-heptafluoro-1-butanol, 1-butanol, 2,2,3,3,4,4,4-heptafluoro, 1h,1h-heptafluorobutanol-1, 1h,1h-heptafluoro-1-butanol, 1,1-dihydroperfluorobutyl alcohol, 2,2,3,3,4,4,4-heptafluorobutanol, 1,1-dihydroperfluorobutanol, 1,1-h,h-heptafluorobutanol, perfluoro-1,1-dihydrobutanol, butanol, 2,2,3,3,4,4,4-heptafluoro
C(C(C(C(F)(F)F)(F)F)(F)F)O
200.056
200.06
2,2,3,3,4,4,4-Heptafluoro-1-butanol
Safety and Handling

Safety and Handling

GHS H Statement
H330-H226-H315-H319-H335
Fatal if inhaled.
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P260-P280h-P305+P351+P338
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.

RTECSNumber : EL4130000

TSCA : TSCA