missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,2,3,3,3-Pentafluoropropylamine, 97%

$129.54 - $444.14

Chemical Identifiers

CAS 422-03-7
Molecular Formula C3H4F5N
Molecular Weight (g/mol) 149.064
MDL Number MFCD00042459
InChI Key DPQNQLKPUVWGHE-UHFFFAOYSA-N
Synonym 2,2,3,3,3-pentafluoropropylamine, pentafluoropropylamine, acmc-1amc1, 1h,1h-pentafluoropropylamine, 2,2,3,3,3-penta-fluoropropylamine, 2,2,3,3,3-pentafluoro-propylamine, 2,2,3,3,3-pentafluoro-propyl amine, 1-propanamine,2,2,3,3,3-pentafluoro, 1-propanamine, 2,2,3,3,3-pentafluoro
PubChem CID 79002
IUPAC Name 2,2,3,3,3-pentafluoropropan-1-amine
SMILES C(C(C(F)(F)F)(F)F)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1681903 View Documents Alfa Aesar™
L1681903
1g Each for $129.54
 
AAL1681906 View Documents Alfa Aesar™
L1681906
5g Each for $444.14
 
Specifications

Chemical Identifiers

422-03-7
149.064
DPQNQLKPUVWGHE-UHFFFAOYSA-N
79002
C(C(C(F)(F)F)(F)F)N
C3H4F5N
MFCD00042459
2,2,3,3,3-pentafluoropropylamine, pentafluoropropylamine, acmc-1amc1, 1h,1h-pentafluoropropylamine, 2,2,3,3,3-penta-fluoropropylamine, 2,2,3,3,3-pentafluoro-propylamine, 2,2,3,3,3-pentafluoro-propyl amine, 1-propanamine,2,2,3,3,3-pentafluoro, 1-propanamine, 2,2,3,3,3-pentafluoro
2,2,3,3,3-pentafluoropropan-1-amine

Specifications

>100°C (212°F)
1g
C3H4F5N
UN2735
2,2,3,3,3-pentafluoropropylamine, pentafluoropropylamine, acmc-1amc1, 1h,1h-pentafluoropropylamine, 2,2,3,3,3-penta-fluoropropylamine, 2,2,3,3,3-pentafluoro-propylamine, 2,2,3,3,3-pentafluoro-propyl amine, 1-propanamine,2,2,3,3,3-pentafluoro, 1-propanamine, 2,2,3,3,3-pentafluoro
C(C(C(F)(F)F)(F)F)N
149.064
149.06
2,2,3,3,3-Pentafluoropropylamine
49°C to 50°C
422-03-7
MFCD00042459
1743135
DPQNQLKPUVWGHE-UHFFFAOYSA-N
2,2,3,3,3-pentafluoropropan-1-amine
79002
97%
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.

Recommended Storage : 2° to 8°C