missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,2,3,3,3-Pentafluoropropyl p-Toluenesulfonate, TCI America™

Manufacturer:  TCI America P183425G

 View more versions of this product

Catalog No. P183425G



Specifications

Specifications

2,2,3,3,3-Pentafluoropropyl p-Toluenesulfonate
White-Yellow
25g
≥98.0% (GC)
MFCD02093346
JBHQQXONFHOEQU-UHFFFAOYSA-N
2,2,3,3,3-pentafluoropropyl 4-methylbenzenesulfonate
2776017
Crystal-Powder at 20°C
75°C
54°C
565-42-4
C10H9F5O3S
p-Toluenesulfonic Acid 2,2,3,3,3-Pentafluoropropyl Ester, 2,2,3,3,3-Pentafluoropropyl Tosylate
CC1=CC=C(C=C1)S(=O)(=O)OCC(C(F)(F)F)(F)F
304.231
304.23

Chemical Identifiers

565-42-4
304.231
JBHQQXONFHOEQU-UHFFFAOYSA-N
2776017
CC1=CC=C(C=C1)S(=O)(=O)OCC(C(F)(F)F)(F)F
C10H9F5O3S
MFCD02093346
p-Toluenesulfonic Acid 2,2,3,3,3-Pentafluoropropyl Ester, 2,2,3,3,3-Pentafluoropropyl Tosylate
2,2,3,3,3-pentafluoropropyl 4-methylbenzenesulfonate
Safety and Handling

Safety and Handling

TSCA : No

Documents