missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2,2,3,3,3-Pentafluoropropyl p-Toluenesulfonate 98.0+%, TCI America™
$293.65 - $889.22
Chemical Identifiers
CAS | 565-42-4 |
---|---|
Molecular Formula | C10H9F5O3S |
Molecular Weight (g/mol) | 304.23 |
MDL Number | MFCD02093346 |
InChI Key | JBHQQXONFHOEQU-UHFFFAOYSA-N |
Synonym | p-Toluenesulfonic Acid 2,2,3,3,3-Pentafluoropropyl Ester, 2,2,3,3,3-Pentafluoropropyl Tosylate |
PubChem CID | 2776017 |
IUPAC Name | 2,2,3,3,3-pentafluoropropyl 4-methylbenzene-1-sulfonate |
SMILES | CC1=CC=C(C=C1)S(=O)(=O)OCC(F)(F)C(F)(F)F |
Chemical Identifiers
565-42-4 | |
304.23 | |
JBHQQXONFHOEQU-UHFFFAOYSA-N | |
2776017 | |
CC1=CC=C(C=C1)S(=O)(=O)OCC(F)(F)C(F)(F)F |
C10H9F5O3S | |
MFCD02093346 | |
p-Toluenesulfonic Acid 2,2,3,3,3-Pentafluoropropyl Ester, 2,2,3,3,3-Pentafluoropropyl Tosylate | |
2,2,3,3,3-pentafluoropropyl 4-methylbenzene-1-sulfonate |
Specifications
565-42-4 | |
White-Yellow | |
C10H9F5O3S | |
5 g | |
JBHQQXONFHOEQU-UHFFFAOYSA-N | |
2,2,3,3,3-pentafluoropropyl 4-methylbenzene-1-sulfonate | |
2776017 | |
≥98.0% (GC) | |
2,2,3,3,3-Pentafluoropropyl p-Toluenesulfonate |
54°C | |
75°C | |
MFCD02093346 | |
p-Toluenesulfonic Acid 2,2,3,3,3-Pentafluoropropyl Ester, 2,2,3,3,3-Pentafluoropropyl Tosylate | |
CC1=CC=C(C=C1)S(=O)(=O)OCC(F)(F)C(F)(F)F | |
304.23 | |
304.23 | |
Crystalline Powder |
Safety and Handling
TSCA : No