Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,2,3,3,3-Pentafluoropropyl methacrylate, 97%, stab.

Chemical Identifiers

Chemical Name or Material 2,2,3,3,3-Pentafluoropropyl methacrylate
CAS 45115-53-5
Name Note stabilized
Molecular Formula C7H7F5O2
Formula Weight 218.12
MDL Number MFCD00039256
UN Number UN3272
Synonym Methacrylic acid 2,2,3,3,3-pentafluoropropyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2434006

  SDS
alfa aesar™
B2434006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2434014

  SDS
alfa aesar™
B2434014
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 218.12
Assay 97%
Boiling Point 54° to 55°C (100mmHg)
Flash Point 29°C (84°F)
Health Hazard 1 Warning
Recommended Storage 2° to 8°C
TSCA TSCA
CAS 45115-53-5
MDL Number MFCD00039256
Name Note stabilized
Synonym Methacrylic acid 2,2,3,3,3-pentafluoropropyl ester
Health Hazard 2 GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
UN Number UN3272
Quantity 5g
Molecular Formula C7H7F5O2
Chemical Name or Material 2,2,3,3,3-Pentafluoropropyl methacrylate
Health Hazard 3 GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
Safety and Handling

Warning

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C