Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,2,3,3,3-Pentafluoropropionamide, 97%

$79.72 - $79.72

Chemical Identifiers

Chemical Name or Material 2,2,3,3,3-Pentafluoropropionamide
CAS 354-76-7
Molecular Formula CF3CF2CONH2
Formula Weight 163.05
MDL Number MFCD00039771
Beilstein 1773386
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2156606

  SDS
alfa aesar™
B2156606
5g Each for $79.72

AAB2156614

  SDS
alfa aesar™
B2156614
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material 2,2,3,3,3-Pentafluoropropionamide
CAS 354-76-7
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
TSCA TSCA
Assay 97%
Melting Point 92° to 97°C
Molecular Formula CF3CF2CONH2
Quantity 5g
Formula Weight 163.05
MDL Number MFCD00039771
Beilstein 1773386

Applications
2,2,3,3,3-Pentafluoropropionamide, is used as a chemical, organic, pharamceutical intermediate.

Solubility
Soluble in methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.