Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,2,3,3,3-Pentafluoro-1-propanol, 98%

$41.42 - $148.23

Chemical Identifiers

Chemical Name or Material 2,2,3,3,3-Pentafluoro-1-propanol
CAS 422-05-9
Molecular Formula CF3CF2CH2OH
Formula Weight 150.05
MDL Number MFCD00004673
Beilstein 1743133
Synonym 1H,1H-Pentafluoro-1-propanol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1184606

 
alfa aesar™
A1184606
5g Each for $41.42

AAA1184614

 
alfa aesar™
A1184614
25g Each for $148.23
Description & Specifications

Specifications

Formula Weight 150.05
Chemical Name or Material 2,2,3,3,3-Pentafluoro-1-propanol
TSCA TSCA
Assay 98%
Boiling Point 80° to 81°C
Molecular Formula CF3CF2CH2OH
Quantity 5g
MDL Number MFCD00004673
CAS 422-05-9
Beilstein 1743133
Synonym 1H,1H-Pentafluoro-1-propanol
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number UB8800000

Applications
2,2,3,3,3-Pentafluoro-1-propanol is used as Pharmaceutical Intermediates. It can be used in agrochemical, pharmaceutical and dyestuff field etc.

Solubility
Soluble in water.21.9 g/L at 25°C

Notes
Incompatible with strong oxidizing agents, Acid chlorides, Acid anhydrides. Keep away from heat.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.