missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,2,3,3,3-Pentafluoro-1-propanol, 98%

Manufacturer:  Alfa Aesar™ A1184614

 View more versions of this product

Catalog No. AAA1184614



Description

Description

Applications
2,2,3,3,3-Pentafluoro-1-propanol is used as Pharmaceutical Intermediates. It can be used in agrochemical, pharmaceutical and dyestuff field etc.

Solubility
Soluble in water.21.9 g/L at 25°C

Notes
Incompatible with strong oxidizing agents, Acid chlorides, Acid anhydrides. Keep away from heat.
Specifications

Specifications

2,2,3,3,3-Pentafluoro-1-propanol
98%
C3H3F5O
MFCD00004673
1H,1H-Pentafluoro-1-propanol
C(C(C(F)(F)F)(F)F)O
150.048
150.05
422-05-9
80°C to 81°C
25g
1743133
PSQZJKGXDGNDFP-UHFFFAOYSA-N
2,2,3,3,3-pentafluoropropan-1-ol
9872

Chemical Identifiers

422-05-9
150.048
PSQZJKGXDGNDFP-UHFFFAOYSA-N
9872
C(C(C(F)(F)F)(F)F)O
C3H3F5O
MFCD00004673
1H,1H-Pentafluoro-1-propanol
2,2,3,3,3-pentafluoropropan-1-ol
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : UB8800000

TSCA : TSCA

SDS Documents