missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,2,2-Trimethylacetamide, 98+%

Manufacturer:  Alfa Aesar™ A1131909

 View more versions of this product

Catalog No. AAA1131909



Specifications

Specifications

2,2,2-Trimethylacetamide
153°C to 157°C
754-10-9
C5H11NO
MFCD00008011
2,2-Dimethylpropionamide; Pivalamide
CC(C)(C)C(=O)N
101.149
101.15
212°C
10g
≥98%
(CH3)3CCONH2
1742147
XIPFMBOWZXULIA-UHFFFAOYSA-N
2,2-dimethylpropanamide
12957

Chemical Identifiers

754-10-9
101.149
XIPFMBOWZXULIA-UHFFFAOYSA-N
12957
CC(C)(C)C(=O)N
C5H11NO
MFCD00008011
2,2-Dimethylpropionamide; Pivalamide
2,2-dimethylpropanamide
Safety and Handling

Safety and Handling

TSCA : TSCA

SDS Documents