Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,2,2-Trimethylacetamide, 98+%

$56.81 - $795.57

Chemical Identifiers

Chemical Name or Material 2,2,2-Trimethylacetamide
CAS 754-10-9
Molecular Formula C5H11NO
Linear Formula (CH3)3CCONH2
Formula Weight 101.15
MDL Number MFCD00008011
Beilstein 1742147
Synonym 2,2-Dimethylpropionamide; Pivalamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1131909

  SDS
alfa aesar™
A1131909
10g Each for $56.81

AAA1131918

  SDS
alfa aesar™
A1131918
50g Each for $183.82

AAA1131930

  SDS
alfa aesar™
A1131930
250g Each for $795.57
Description & Specifications

Specifications

Formula Weight 101.15
Chemical Name or Material 2,2,2-Trimethylacetamide
TSCA TSCA
Assay ≥98%
Molecular Formula C5H11NO
Linear Formula (CH3)3CCONH2
MDL Number MFCD00008011
CAS 754-10-9
Beilstein 1742147
Synonym 2,2-Dimethylpropionamide; Pivalamide
Boiling Point 212°C
Melting Point 153° to 157°C
Quantity 10g