Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,2,2-Trimethoxy-4,5-dimethyl-1,3,2-dioxaphospholene, 97%

$84.82 - $84.82

Chemical Identifiers

Chemical Name or Material 2,2,2-Trimethoxy-4,5-dimethyl-1,3,2-dioxaphospholene
CAS 1665-79-8
Molecular Formula C7H15O5P
Formula Weight 210.17
MDL Number MFCD00014510
Synonym 2,3-Dihydro-2,2,2-trimethoxy-4,5-dimethyl-1,3,2-dioxaphophole
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2273009

  SDS
alfa aesar™
B2273009
10g Each for $84.82
Description & Specifications

Specifications

Formula Weight 210.17
Chemical Name or Material 2,2,2-Trimethoxy-4,5-dimethyl-1,3,2-dioxaphospholene
TSCA TSCA
Assay 97%
Molecular Formula C7H15O5P
MDL Number MFCD00014510
CAS 1665-79-8
Synonym 2,3-Dihydro-2,2,2-trimethoxy-4,5-dimethyl-1,3,2-dioxaphophole
Boiling Point 64° to 66°C (1mmHg)
Quantity 10g

Applications
2,2,2-Trimethoxy-4,5-dimethyl-1,3,2-dioxaphospholene is used as an organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.