Learn More
2,2,2-Trifluoroethylhydrazine, 70 wt% aqueous solution, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 365760050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
2, 2, 2-Trifluoroethylhydrazine | |
5042-30-8,7732-18-5 | |
75.0,35.0 | |
42°C | |
C2H5F3N2 | |
CF3CH2NHNH2 | |
5g | |
2,2,2-trifluoroethyl hydrazine, 2,2,2-trifluoroethylhydrazine, 70 % in aqueous solution, 2,2,2-trifluoroethyl hydrazine in water, sntljqhjptadtauup@, acmc-20aoks, trifluoroethyl hydrazine, cf3ch2nhnh2, 2,2,2-trifluorethylhydrazine | |
C(C(F)(F)F)NN | |
114.07 | |
114.07 |
70 wt% aqueous solution | |
65.0,25.0 | |
1.2940g/mL | |
Glass bottle | |
1.3600 to 1.364 | |
MFCD00007622 | |
1.294 | |
OPMFFAOEPFATTG-UHFFFAOYSA-N | |
2,2,2-trifluoroethylhydrazine | |
78740 | |
≥65% |
Chemical Identifiers
5042-30-8 | |
114.07 | |
OPMFFAOEPFATTG-UHFFFAOYSA-N | |
78740 | |
C(C(F)(F)F)NN |
C2H5F3N2 | |
MFCD00007622 | |
2,2,2-trifluoroethyl hydrazine, 2,2,2-trifluoroethylhydrazine, 70 % in aqueous solution, 2,2,2-trifluoroethyl hydrazine in water, sntljqhjptadtauup@, acmc-20aoks, trifluoroethyl hydrazine, cf3ch2nhnh2, 2,2,2-trifluorethylhydrazine | |
2,2,2-trifluoroethylhydrazine |
Safety and Handling
GHS H Statement
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Flammable liquid and vapor.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Call a POISON CENTER or
GHS Signal Word: Warning
EINECSNumber : 225-734-
RUO â Research Use Only