Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,2,2-Trifluoroethyl vinyl ether, 97%, stab.

Chemical Identifiers

Chemical Name or Material 2,2,2-Trifluoroethyl vinyl ether
CAS 406-90-6
Name Note stabilized
Molecular Formula C4H5F3O
Linear Formula H2C=CHOCH2CF3
Formula Weight 126.08
MDL Number MFCD00864407
UN Number UN3271
Beilstein 1745876
Merck Index 14,4202
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL2021803

  SDS
alfa aesar™
L2021803
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 126.08
Chemical Name or Material 2,2,2-Trifluoroethyl vinyl ether
MDL Number MFCD00864407
Name Note stabilized
Odor Ether-like
Beilstein 1745876
Linear Formula H2C=CHOCH2CF3
Molecular Formula C4H5F3O
Recommended Storage 2° to 8°C
Quantity 1g
Synonym Fluroxene
RTECS Number KO4250000
Health Hazard 3 GHS P Statement
P210-P280-P281-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Boiling Point 42° to 43°C
Flash Point −30°C (−22°F)
TSCA TSCA
Merck Index 14,4202
UN Number UN3271
Assay 97%
Health Hazard 2 GHS H Statement
H225-H341
Highly flammable liquid and vapor.
Suspected of causing genetic defects.
Health Hazard 1 Danger
CAS 406-90-6
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHERS, N.O.S.
Safety and Handling

Danger

GHS P Statement
P210-P280-P281-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H341
Highly flammable liquid and vapor.
Suspected of causing genetic defects.

2° to 8°C