Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,2,2-Trifluoroethyl methyl ether, 99+%

Chemical Identifiers

Chemical Name or Material 2,2,2-Trifluoroethyl methyl ether
CAS 460-43-5
Molecular Formula C3H5F3O
Linear Formula CH3OCH2CF3
Formula Weight 114.07
MDL Number MFCD00236114
UN Number UN1993
Beilstein 1734776
Synonym Methyl 2,2,2-trifluoroethyl ether
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1788706

  SDS
alfa aesar™
L1788706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1788714

  SDS
alfa aesar™
L1788714
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 114.07
CAS 460-43-5
Molecular Formula C3H5F3O
Health Hazard 3 GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Boiling Point 30°C
Health Hazard 1 Danger
Assay ≥99%
Odor Ether-like
UN Number UN1993
Flash Point −8°C (−18°F)
Chemical Name or Material 2,2,2-Trifluoroethyl methyl ether
MDL Number MFCD00236114
Recommended Storage 2° to 8°C
Beilstein 1734776
Linear Formula CH3OCH2CF3
DOT Information Transport Hazard Class: 3; Packing Group: I; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
Quantity 5g
Health Hazard 2 GHS H Statement
H224
Extremely flammable liquid and vapor.
Synonym Methyl 2,2,2-trifluoroethyl ether
Safety and Handling

Danger

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H224
Extremely flammable liquid and vapor.

2° to 8°C