missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,2,2-Trifluoroethyl Methacrylate (stabilized with MEHQ), TCI America™

$36.67 - $449.17

Chemical Identifiers

CAS 352-87-4
Molecular Formula C6H7F3O2
Molecular Weight (g/mol) 168.115
MDL Number MFCD00013576
InChI Key QTKPMCIBUROOGY-UHFFFAOYSA-N
Synonym 2,2,2-trifluoroethyl methacrylate, trifluoroethyl methacrylate, methacrylic acid 2,2,2-trifluoroethyl ester, 2,2,2-trifluoroethylmethacrylate, 2-propenoic acid, 2-methyl-, 2,2,2-trifluoroethyl ester, 2-methyl-acrylic acid 2,2,2-trifluoro-ethyl ester, 2-propenoic acid, 2-methyl-, trifluoroethyl ester, 2-methyl-2-propenoic acid 2,2,2-trifluoroethyl ester, 2,2,2-trifluoroethyl m, acmc-1af8z
PubChem CID 9608
IUPAC Name 2,2,2-trifluoroethyl 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OCC(F)(F)F
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M073825ML View Documents TCI America
M073825ML
25mL Each for $36.67
 
M0738100ML View Documents TCI America
M0738100ML
100mL Each for $128.34
 
M0738500ML View Documents TCI America
M0738500ML
500mL Each for $449.17
 
Specifications

Chemical Identifiers

352-87-4
168.115
QTKPMCIBUROOGY-UHFFFAOYSA-N
9608
CC(=C)C(=O)OCC(F)(F)F
C6H7F3O2
MFCD00013576
2,2,2-trifluoroethyl methacrylate, trifluoroethyl methacrylate, methacrylic acid 2,2,2-trifluoroethyl ester, 2,2,2-trifluoroethylmethacrylate, 2-propenoic acid, 2-methyl-, 2,2,2-trifluoroethyl ester, 2-methyl-acrylic acid 2,2,2-trifluoro-ethyl ester, 2-propenoic acid, 2-methyl-, trifluoroethyl ester, 2-methyl-2-propenoic acid 2,2,2-trifluoroethyl ester, 2,2,2-trifluoroethyl m, acmc-1af8z
2,2,2-trifluoroethyl 2-methylprop-2-enoate

Specifications

102°C
25mL
C6H7F3O2
3272
QTKPMCIBUROOGY-UHFFFAOYSA-N
2,2,2-trifluoroethyl 2-methylprop-2-enoate
9608
Clear Liquid at 20°C
2,2,2-Trifluoroethyl Methacrylate (stabilized with MEHQ)
Colorless
352-87-4
MFCD00013576
2,2,2-trifluoroethyl methacrylate, trifluoroethyl methacrylate, methacrylic acid 2,2,2-trifluoroethyl ester, 2,2,2-trifluoroethylmethacrylate, 2-propenoic acid, 2-methyl-, 2,2,2-trifluoroethyl ester, 2-methyl-acrylic acid 2,2,2-trifluoro-ethyl ester, 2-propenoic acid, 2-methyl-, trifluoroethyl ester, 2-methyl-2-propenoic acid 2,2,2-trifluoroethyl ester, 2,2,2-trifluoroethyl m, acmc-1af8z
CC(=C)C(=O)OCC(F)(F)F
168.115
168.12
≥98.0% (GC)
Safety and Handling

Safety and Handling

EINECSNumber : (2)-1031

TSCA : Yes