Please login to your online account to display your discounted pricing

2,2,2-Trifluoroethyl methacrylate, 99%, stabilized, ACROS Organics™


Quantity 5g

Manufacturer: thermo fisher scientific  312780050

 View more versions of this product

Catalog No. AC312780050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5% (GC)
Physical Form Crystals or Crystalline Powder
Formula Weight 168.12
Quantity 5g
Color White
TSCA TSCA
EINECS Number 206-525-3
Density 1.18g/mL
CAS Max % 100.0
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS Min % 98.5
Refractive Index 1.3600 to 1.3620 (20°C, 589nm)
Infrared Spectrum Authentic
Stabilizer 100ppm MEHQ
MDL Number MFCD00013576
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Highly flammable liquid and vapor.
Chemical Name or Material 2, 2, 2-Trifluoroethyl methacrylate
Name Note 99%
Molecular Formula C6H7F3O2
CAS 352-87-4
Linear Formula HC=C(CH3)COCHCF3
Assay Percent Range 99%

Specifications

Chemical Name or Material 2, 2, 2-Trifluoroethyl methacrylate
Name Note 99%
CAS 352-87-4
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5% (GC)
Assay Percent Range 99%
Molecular Formula C6H7F3O2
Linear Formula HC=C(CH3)COCHCF3
Formula Weight 168.12
MDL Number MFCD00013576
Accessories

GHS Signal Word: Danger

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Highly flammable liquid and vapor.

SDS & Safety Documents