missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,2,2-Trifluoroethyl methacrylate, 99%, stabilized, ACROS Organics™

Chemical Identifiers

CAS 352-87-4
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Molecular Formula C6H7F3O2
Linear Formula HC=C(CH3)COCHCF3
Formula Weight 168.12
MDL Number MFCD00013576
Chemical Name or Material 2, 2, 2-Trifluoroethyl methacrylate, 99%
Assay ≥98.5% (GC)
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC312780050 SDS Thermo Fisher Scientific
312780050
5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC312780250 SDS Thermo Fisher Scientific
312780250
Glass bottle 25g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

352-87-4
100.0
C6H7F3O2
168.12
2, 2, 2-Trifluoroethyl methacrylate, 99%
98.5
99%
HC=C(CH3)COCHCF3
MFCD00013576
≥98.5% (GC)

Specifications

1.18g/mL
5g
98.5
Authentic
100ppm MEHQ
99%
HC=C(CH3)COCHCF3
MFCD00013576
≥98.5% (GC)
White
352-87-4
100.0
1.3600 to 1.3620 (20°C, 589nm)
Crystals or Crystalline Powder
C6H7F3O2
168.12
2, 2, 2-Trifluoroethyl methacrylate, 99%

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Highly flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 206-525-3

TSCA : TSCA