Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,2,2-Trifluoroethyl methacrylate, 98%, stab. with 30-50 ppm 4-methoxyphenol

Chemical Identifiers

Chemical Name or Material 2,2,2-Trifluoroethyl methacrylate
CAS 352-87-4
Name Note stabilized with 30 to 50 ppm 4-methoxyphenol
Molecular Formula C6H7F3O2
Formula Weight 168.12
MDL Number MFCD00013576
UN Number UN3272
Beilstein 4175872
Synonym Methacrylic acid 2,2,2-trifluoroethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1701214

  SDS
alfa aesar™
A1701214
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 168.12
Name Note stabilized with 30 to 50 ppm 4-methoxyphenol
Boiling Point 29° to 30°C (40mmHg)
Beilstein 4175872
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 25g
Assay 98%
UN Number UN3272
CAS 352-87-4
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
Recommended Storage 2° to 8°C
Molecular Formula C6H7F3O2
Flash Point 25°C (77°F)
Synonym Methacrylic acid 2,2,2-trifluoroethyl ester
MDL Number MFCD00013576
Chemical Name or Material 2,2,2-Trifluoroethyl methacrylate
Health Hazard 3 GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
TSCA TSCA
Odor Pungent
Safety and Handling

Warning

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C