Please login to your online account to display your discounted pricing

2,2,2-Trifluoroethyl formate ≥96%, ACROS Organics™

$42.23 - $42.23

Chemical Identifiers

Chemical Name or Material 2, 2, 2-Trifluoroethyl formate
CAS 32042-38-9
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C3H3F3O2
Linear Formula CF3CH2OC(O)H
Formula Weight 128.05
MDL Number MFCD06797733
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC390380010

  SDS
thermo fisher scientific
AC390380010
Glass bottle 1g Each for $42.23
Description & Specifications

Specifications

Assay ≥94%
Physical Form Liquid
Formula Weight 128.05
Quantity 1g
Color Clear colorless to light yellow
Density 1.3170g/mL
CAS Max % 100.0
CAS Min % 94.0
CAS 32042-38-9
MDL Number MFCD06797733
Packaging Glass bottle
Infrared Spectrum Authentic
Linear Formula CF3CH2OC(O)H
Molecular Formula C3H3F3O2
Chemical Name or Material 2, 2, 2-Trifluoroethyl formate
Assay Percent Range 95%
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Causes serious eye irritation.
May cause respiratory irritation.
Highly flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Causes serious eye irritation.
May cause respiratory irritation.
Highly flammable liquid and vapor.