Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,2,2-Trifluoroethyl acetate, 97%

$105.03 - $105.03

Chemical Identifiers

Chemical Name or Material 2,2,2-Trifluoroethyl acetate
CAS 406-95-1
Molecular Formula C4H5F3O2
Linear Formula CH3CO2CH2CF3
Formula Weight 142.08
MDL Number MFCD00198018
UN Number UN3272
Beilstein 1757502
Synonym Acetic acid 2,2,2-trifluoroethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1687114

  SDS
alfa aesar™
L1687114*
25g Each for $105.03

AAL1687122

  SDS
alfa aesar™
L1687122
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 142.08
Chemical Name or Material 2,2,2-Trifluoroethyl acetate
Assay 97%
Molecular Formula C4H5F3O2
Linear Formula CH3CO2CH2CF3
MDL Number MFCD00198018
CAS 406-95-1
UN Number UN3272
Beilstein 1757502
Synonym Acetic acid 2,2,2-trifluoroethyl ester
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H225
Highly flammable liquid and vapor.
Health Hazard 3 GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ESTERS, N.O.S.
Flash Point 11°C (51°F)
Boiling Point 78°C
Quantity 25g

Applications
2,2,2-Trifluoroethyl trifluoroacetate is used in the preparation of dimethyl (3,3,3-trifluoro-2,2-dihydroxypropyl)phosphonate

Solubility
Soluble in water (16 g/L) (25°C)

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Safety and Handling

Danger

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225
Highly flammable liquid and vapor.