Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,2,2-Trifluoroethanol, NMR grade, 99.5%


Quantity 25g

Manufacturer: alfa aesar™  L1689814

 View more versions of this product

Catalog No. AAL1689814

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 100.04
Molecular Formula C2H3F3O
CAS 75-89-8
Chemical Name or Material 2,2,2-Trifluoroethanol
Synonym Trifluoroethyl alcohol
Linear Formula CF3CH2OH
Beilstein 1733203
RTECS Number KM5250000
Purity Grade NMR Grade
TSCA TSCA
MDL Number MFCD00004672
Health Hazard 1 Danger
Assay 99.5%
UN Number UN1992
Melting Point −44°C
Health Hazard 2 GHS H Statement
H301-H311-H318-H226-H332-H315-H335
Toxic if swallowed.
Toxic in contact with skin.
Causes serious eye damage.
Flammable liquid and vapor.
Harmful if inhaled.
Causes skin irritation.
May cause respiratory irritation.
Boiling Point 74° to 75°C
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, TOXIC, N.O.S.
Health Hazard 3 GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Flash Point 29°C (84°F)
Quantity 25g

Specifications

Chemical Name or Material 2,2,2-Trifluoroethanol
CAS 75-89-8
Purity Grade NMR Grade
Molecular Formula C2H3F3O
Linear Formula CF3CH2OH
Formula Weight 100.04
MDL Number MFCD00004672
UN Number UN1992
Beilstein 1733203
Synonym Trifluoroethyl alcohol
Accessories

Danger

GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H311-H318-H226-H332-H315-H335
Toxic if swallowed.
Toxic in contact with skin.
Causes serious eye damage.
Flammable liquid and vapor.
Harmful if inhaled.
Causes skin irritation.
May cause respiratory irritation.

SDS & Safety Documents