Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,2,2-Trifluoroethanol-d3, 99% (Isotopic)

NMR Solvent

Chemical Identifiers

Chemical Name or Material 2,2,2-Trifluoroethanol-d3
CAS 77253-67-9
Name Note Isotopic
Formula Weight 103.06
MDL Number MFCD00037682
UN Number UN1992
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4236346

  SDS
alfa aesar™
4236346
5 x 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4236303

  SDS
alfa aesar™
4236303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material 2,2,2-Trifluoroethanol-d3
Name Note Isotopic
CAS 77253-67-9
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H331-H318-H226-H302-H312-H315-H335
Toxic if inhaled.
Causes serious eye damage.
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Packaging Prescored Ampules
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, TOXIC, N.O.S.
Sensitivity Hygroscopic
Density 1.45g/mL
Boiling Point 77° to 80°C
Melting Point −44°C
Physical Form Liquid
Assay 99%
Quantity 5 x 1g
Formula Weight 103.06
MDL Number MFCD00037682
UN Number UN1992
Safety and Handling

Danger

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H318-H226-H302-H312-H315-H335
Toxic if inhaled.
Causes serious eye damage.
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
May cause respiratory irritation.