missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,2,2-Trifluoroethanol-d2, 98% (Isotopic)

NMR Solvent

Chemical Identifiers

CAS 132248-58-9
Formula Weight 102.02
MDL Number MFCD00082609
UN Number UN1992
Chemical Name or Material 2,2,2-Trifluoroethanol-d2, Isotopic
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA4236203 SDS Alfa Aesar™
4236203
1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

132248-58-9
MFCD00082609
2,2,2-Trifluoroethanol-d2, Isotopic
102.02
UN1992

Specifications

132248-58-9
77° to 80°C
Liquid
102.02
UN1992
98%
Hygroscopic
−44°C
1g
MFCD00082609
2,2,2-Trifluoroethanol-d2, Isotopic

Safety and Handling

GHS H Statement
H331-H318-H226-H302-H312-H315-H335
Toxic if inhaled.
Causes serious eye damage.
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, TOXIC, N.O.S.