missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Honeywell Fluka™ 2,2,2-Trifluoroethanol, ≥99.8% (GC), Eluent additive for LC-MS, Honeywell™ Fluka™

Eluent additive for LC-MS

$414.83 - $414.83

Chemical Identifiers

CAS 75-89-8
Molecular Formula C2H3F3O
Molecular Weight (g/mol) 100.04
MDL Number MFCD00004672
InChI Key RHQDFWAXVIIEBN-UHFFFAOYSA-N
Synonym trifluoroethanol, ethanol, 2,2,2-trifluoro, 2,2,2-trifluoroethan-1-ol, fluorinol 85, 2,2,2-trifluoroethyl alcohol, trifluoroethyl alcohol, trifluoro ethanol, tfe, perfluoro-1,1-dihydroethanol, tfetoh
PubChem CID 6409
ChEBI CHEBI:42330
IUPAC Name 2,2,2-trifluoroethanol
SMILES C(C(F)(F)F)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
60044574 View Documents Honeywell Fluka™
0584150ML
50mL Glass Bottle Each for $414.83
 
Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Chemical Identifiers

75-89-8
100.04
RHQDFWAXVIIEBN-UHFFFAOYSA-N
6409
2,2,2-trifluoroethanol
C2H3F3O
MFCD00004672
trifluoroethanol, ethanol, 2,2,2-trifluoro, 2,2,2-trifluoroethan-1-ol, fluorinol 85, 2,2,2-trifluoroethyl alcohol, trifluoroethyl alcohol, trifluoro ethanol, tfe, perfluoro-1,1-dihydroethanol, tfetoh
CHEBI:42330
C(C(F)(F)F)O

Specifications

1.391g/mL at 20°C, 1.373g/mL at 25°C
Glass Bottle
3.5 (vs air)
77°C to 80°C
-44°C (literature)
75-89-8
CF3CH2OH
UN1986
trifluoroethanol, ethanol, 2,2,2-trifluoro, 2,2,2-trifluoroethan-1-ol, fluorinol 85, 2,2,2-trifluoroethyl alcohol, trifluoroethyl alcohol, trifluoro ethanol, tfe, perfluoro-1,1-dihydroethanol, tfetoh
C(C(F)(F)F)O
100.04
CHEBI:42330
2,2,2-Trifluoroethanol
29°C (84.2°F)
n20/D 1.3(lit.)
70mmHg (25°C)
Colorless
50mL
C2H3F3O
MFCD00004672
1733203
RHQDFWAXVIIEBN-UHFFFAOYSA-N
2,2,2-trifluoroethanol
6409
100.04g/mol
Safety and Handling

Safety and Handling

P261-P280-P301 + P310-P305 + P351 + P338-P311

H226-H301 + H331-H312-H315-H318-H335

DOTInformation : 6.1(3)/PGIII

EINECSNumber : 200-913-6

RTECSNumber : KM5250000