Learn More
2,2,2-Trifluoroacetophenone, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1140306
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2,2,2-Trifluoroacetophenone | |
-40°C | |
165°C to 166°C | |
C8H5F3O | |
MFCD00000420 | |
UN1224 | |
trifluoroacetophenone, 2,2,2-trifluoroacetophenone, 2,2,2-trifluoro-1-phenylethan-1-one, phenyl trifluoromethyl ketone, trifluoromethyl phenyl ketone, alpha,alpha,alpha-trifluoroacetophenone, ethanone, 2,2,2-trifluoro-1-phenyl, 2,2,2-trifluoro-1-phenyl-ethanone, trifluoroacetylbenzene, unii-6t7l1upy09 | |
C1=CC=C(C=C1)C(=O)C(F)(F)F | |
174.122 | |
174.12 |
434-45-7 | |
1.24 | |
42°C (108°F) | |
1.4595 | |
5 g | |
1866286 | |
KZJRKRQSDZGHEC-UHFFFAOYSA-N | |
2,2,2-trifluoro-1-phenylethanone | |
9905 | |
98% |
Chemical Identifiers
434-45-7 | |
174.122 | |
KZJRKRQSDZGHEC-UHFFFAOYSA-N | |
9905 | |
C1=CC=C(C=C1)C(=O)C(F)(F)F |
C8H5F3O | |
MFCD00000420 | |
trifluoroacetophenone, 2,2,2-trifluoroacetophenone, 2,2,2-trifluoro-1-phenylethan-1-one, phenyl trifluoromethyl ketone, trifluoromethyl phenyl ketone, alpha,alpha,alpha-trifluoroacetophenone, ethanone, 2,2,2-trifluoro-1-phenyl, 2,2,2-trifluoro-1-phenyl-ethanone, trifluoroacetylbenzene, unii-6t7l1upy09 | |
2,2,2-trifluoro-1-phenylethanone |
Safety and Handling
GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H226-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: KETONES, LIQUID, N.O.S.
EINECSNumber : 207-103-1
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only