missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,2,2-Trifluoro-N-phenylacetimidoyl Chloride, TCI America™

Manufacturer:  TCI America T254725G

 View more versions of this product

Catalog No. T254725G



Specifications

Specifications

2,2,2-Trifluoro-N-phenylacetimidoyl Chloride
Yellow
61881-19-4
2924
C1=CC=C(C=C1)N=C(C(F)(F)F)Cl
207.58
207.58
≥98.0% (GC,T)
53°C
25g
C8H5ClF3N
UKKALSJSKAHGTL-UHFFFAOYSA-N
2,2,2-trifluoro-N-phenylethanimidoyl chloride
10058869
Clear Liquid at 20°C

Chemical Identifiers

61881-19-4
207.58
10058869
C1=CC=C(C=C1)N=C(C(F)(F)F)Cl
C8H5ClF3N
UKKALSJSKAHGTL-UHFFFAOYSA-N
2,2,2-trifluoro-N-phenylethanimidoyl chloride
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

Documents