missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,2,2-Trifluoro-N,N-dimethylacetamide, TCI America™

Manufacturer:  TCI America T32625G

 View more versions of this product

Catalog No. T32625G



Specifications

Specifications

2,2,2-Trifluoro-N,N-dimethylacetamide
Colorless
1547-87-1
MFCD00043555
WXBWKMLIVXELSF-UHFFFAOYSA-N
2,2,2-trifluoro-N,N-dimethylacetamide
350491
Clear Liquid at 20°C
136°C
5g
C4H6F3NO
1993
CN(C)C(=O)C(F)(F)F
141.093
141.09
≥98.0% (GC)

Chemical Identifiers

1547-87-1
141.093
WXBWKMLIVXELSF-UHFFFAOYSA-N
2,2,2-trifluoro-N,N-dimethylacetamide
C4H6F3NO
MFCD00043555
350491
CN(C)C(=O)C(F)(F)F
Safety and Handling

Safety and Handling

TSCA : No

Documents