Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,2,2-Trifluoro-1-phenylethanol, 98%

$83.93 - $337.51

Chemical Identifiers

Chemical Name or Material 2,2,2-Trifluoro-1-phenylethanol
CAS 340-05-6
Molecular Formula C8H7F3O
Formula Weight 176.14
MDL Number MFCD00004498
Beilstein 3197623
Synonym alpha-(Trifluoromethyl)benzyl alcohol; 1-Phenyl-2,2,2-trifluoroethanol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2123506

  SDS
alfa aesar™
B2123506
5g Each for $83.93

AAB2123514

  SDS
alfa aesar™
B2123514
25g Each for $337.51
Description & Specifications

Specifications

Formula Weight 176.14
Chemical Name or Material 2,2,2-Trifluoro-1-phenylethanol
Health Hazard 1 Warning
MDL Number MFCD00004498
Synonym alpha-(Trifluoromethyl)benzyl alcohol; 1-Phenyl-2,2,2-trifluoroethanol
Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 5g
Molecular Formula C8H7F3O
Flash Point 84°C (184°F)
Beilstein 3197623
Assay 98%
Health Hazard 2 GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Boiling Point 57° to 59°C (2mmHg)
CAS 340-05-6
Safety and Handling

Warning

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.