Learn More
2,2,2-Trichloroethyl chloroformate, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 161811000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Coupling reagentSpecifications
2, 2, 2-Trichloroethyl chloroformate | |
17341-93-4 | |
100.0 | |
Yellow | |
171°C to 172°C | |
Authentic | |
Glass bottle | |
1.4690 to 1.471 | |
MFCD00000810 | |
02,426; 03,296; 04,522; 05,686; 07,383 | |
2,2,2-trichloroethyl chloroformate, trichloroethyl chloroformate, 2,2,2-trichloroethylchloroformate, carbonochloridic acid, 2,2,2-trichloroethyl ester, 2,2,2-trichloroethoxycarbonyl chloride, chloroformic acid 2,2,2-trichloroethyl ester, formic acid, chloro-, 2,2,2-trichloroethyl ester, chloro 2,2,2-trichloroethoxy methanone, beta, beta, beta-trichloroethoxycarbonyl chloride, troc | |
LJCZNYWLQZZIOS-UHFFFAOYSA-N | |
2,2,2-trichloroethyl carbonochloridate | |
87063 | |
97% |
96% | |
95.0 | |
1°C | |
1.5390g/mL | |
>100°C | |
96% min. (GC) | |
C3H2Cl4O2 | |
ClCO2CH2CCl3 | |
100 g | |
1.539 | |
Solubility in water: decomposes. Other solubilities: soluble in most common organic solvents | |
C(C(Cl)(Cl)Cl)OC(=O)Cl | |
211.86 | |
211.86 | |
Liquid |
Chemical Identifiers
17341-93-4 | |
LJCZNYWLQZZIOS-UHFFFAOYSA-N | |
87063 | |
C(C(Cl)(Cl)Cl)OC(=O)Cl |
MFCD00000810 | |
2,2,2-trichloroethyl chloroformate, trichloroethyl chloroformate, 2,2,2-trichloroethylchloroformate, carbonochloridic acid, 2,2,2-trichloroethyl ester, 2,2,2-trichloroethoxycarbonyl chloride, chloroformic acid 2,2,2-trichloroethyl ester, formic acid, chloro-, 2,2,2-trichloroethyl ester, chloro 2,2,2-trichloroethoxy methanone, beta, beta, beta-trichloroethoxycarbonyl chloride, troc | |
2,2,2-trichloroethyl carbonochloridate |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Store in a dry place. Store in a closed container.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 241-363-7
TSCA : TSCA
RUO – Research Use Only