missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,2,2-Trichloroethanol, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 139445000

 View more versions of this product

Catalog No. AC139445000




Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2, 2, 2-Trichloroethanol
115-20-8
100.0
Colorless to Yellow
151.0°C
Authentic
Glass bottle
1.4885 to 1.4905
MFCD00004677
01, 338
1.55
trichloroethanol, trichlorethanol, trichloroethyl alcohol, ethanol, 2,2,2-trichloro, 2,2,2-trichloro-1-ethanol, hydroxymethyl trichloromethane, 2,2,2-trichloroethyl alcohol, 2,2,2-trichloroethan-1-ol, unii-aw835aj62n, beta-trichloroethanol
KPWDGTGXUYRARH-UHFFFAOYSA-N
2,2,2-trichloroethanol
8259
149.4
Liquid
99%
98.5
17.8°C
1.5500g/mL
88°C
98.5% min. (GC)
C2H3Cl3O
CCl3CH2OH
500 g
03,295; 04,521; 06,605
15, 9803
Solubility in water: slightly soluble. Other solubilities: miscible with alcohol,ether and carbon,tetrachloride
C(C(Cl)(Cl)Cl)O
149.4
CHEBI:28094
99%

Chemical Identifiers

115-20-8
149.4
KPWDGTGXUYRARH-UHFFFAOYSA-N
8259
2,2,2-trichloroethanol
C2H3Cl3O
MFCD00004677
trichloroethanol, trichlorethanol, trichloroethyl alcohol, ethanol, 2,2,2-trichloro, 2,2,2-trichloro-1-ethanol, hydroxymethyl trichloromethane, 2,2,2-trichloroethyl alcohol, 2,2,2-trichloroethan-1-ol, unii-aw835aj62n, beta-trichloroethanol
CHEBI:28094
C(C(Cl)(Cl)Cl)O
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes serious eye damage.
Causes skin irritation.
May cause drowsiness or dizziness.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Wash face,hands and any exposed skin thoroughly after handling.
Do not eat,drink or smoke when using this product.
Rinse mouth.
IF IN EY

GHS Signal Word: Danger

EINECSNumber : 204-071-0

RTECSNumber : KM3850000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only