Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,2,2-Trichloroacetamide, 99%

$159.04 - $159.04

Chemical Identifiers

Chemical Name or Material 2,2,2-Trichloroacetamide
CAS 594-65-0
Molecular Formula Cl3CCONH2
Formula Weight 162.40g/mol
MDL Number MFCD00008009
Beilstein 1754028
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0041118

 
alfa aesar™
L00411-18
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0041130

 
alfa aesar™
L00411-30
250g Each for $159.04
Description & Specifications

Specifications

Melting Point 140° to 144°C
Boiling Point 238° to 240°C
Quantity 50g
Chemical Name or Material 2,2,2-Trichloroacetamide
CAS 594-65-0
Assay 99%
Molecular Formula Cl3CCONH2
Formula Weight 162.40g/mol
MDL Number MFCD00008009
Beilstein 1754028
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number AC9275000
TSCA TSCA
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.