Please login to your online account to display your discounted pricing

2,2,2-Trichloro-1-ethoxyethanol, 85%, Tech., ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC346531000

 View more versions of this product

Catalog No. AC346531000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 193.46
Purity Grade Technical Grade
Physical Form Crystalline Powder, Crystals, Needles or Low Melting Mass
Quantity 100g
Color White to light yellow
Merck Index 15, 2069
CAS Min % 80.0
CAS Max % 100.0
Infrared Spectrum Authentic
MDL Number MFCD00013668
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Chemical Name or Material 2, 2, 2-Trichloro-1-ethoxyethanol
CAS 515-83-3
Molecular Formula C4H7Cl3O2
Assay Percent Range 85%
Packaging Glass bottle
Water 3% max. (K.F.)
Synonym Chloral alcoholate
NMR 80% min.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 2, 2, 2-Trichloro-1-ethoxyethanol
CAS 515-83-3
CAS Min % 80.0
CAS Max % 100.0
Purity Grade Technical Grade
Assay Percent Range 85%
Molecular Formula C4H7Cl3O2
Formula Weight 193.46
MDL Number MFCD00013668
Accessories

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.

SDS & Safety Documents