Please login to your online account to display your discounted pricing

2,2,2-Trichloro-1-ethoxyethanol, 85%, Tech., ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2, 2, 2-Trichloro-1-ethoxyethanol
CAS 515-83-3
CAS Min % 80.0
CAS Max % 100.0
Purity Grade Technical Grade
Assay Percent Range 85%
Molecular Formula C4H7Cl3O2
Formula Weight 193.46
MDL Number MFCD00013668
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC346531000

  SDS
thermo fisher scientific
AC346531000
Glass bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 193.46
Purity Grade Technical Grade
Physical Form Crystalline Powder, Crystals, Needles or Low Melting Mass
Quantity 100g
Color White to light yellow
Merck Index 15, 2069
CAS Min % 80.0
CAS Max % 100.0
Infrared Spectrum Authentic
MDL Number MFCD00013668
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Chemical Name or Material 2, 2, 2-Trichloro-1-ethoxyethanol
CAS 515-83-3
Molecular Formula C4H7Cl3O2
Assay Percent Range 85%
Packaging Glass bottle
Water 3% max. (K.F.)
Synonym Chloral alcoholate
NMR 80% min.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.