Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,2,2-Trichloro-1,1-bis(trifluoromethyl)ethanol, 97%

$83.18 - $281.77

Chemical Identifiers

Chemical Name or Material 2,2,2-Trichloro-1,1-bis(trifluoromethyl)ethanol
CAS 7594-49-2
Molecular Formula C4HCl3F6O
Formula Weight 285.4
MDL Number MFCD00236021
Beilstein 1841641
Synonym 1,1-Bis(trifluoromethyl)-2,2,2-trichloroethanol; 1,1,1,3,3,3-Hexafluoro-2-trichloromethyl-2-propanol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1379203

  SDS
alfa aesar™
L1379203
1g Each for $83.18

AAL1379206

  SDS
alfa aesar™
L1379206
5g Each for $281.77
Description & Specifications

Specifications

Formula Weight 285.4
Chemical Name or Material 2,2,2-Trichloro-1,1-bis(trifluoromethyl)ethanol
CAS 7594-49-2
MDL Number MFCD00236021
Health Hazard 1 Danger
Beilstein 1841641
Synonym 1,1-Bis(trifluoromethyl)-2,2,2-trichloroethanol; 1,1,1,3,3,3-Hexafluoro-2-trichloromethyl-2-propanol
Health Hazard 2 GHS H Statement
H301-H332-H315-H319-H335
Toxic if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Molecular Formula C4HCl3F6O
Assay 97%
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage 2° to 8°C
Quantity 1g
Boiling Point 139°C
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H332-H315-H319-H335
Toxic if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C